All Photos(1)
GE19-5006-01
Sigma-Aldrich
AC 10 Flow Adapter
NACRES:
SB.52
application(s)
LPLC: suitable
Related Categories
General description
The basic C columns are of fixed volume but can be upgraded by adding flow adapters (which keep bed height constant and protect from insoluble particles) and a thermostatic jacket. Three adjustable flow adapters are available for the C column series (AC 10, AC 16, and AC 26), allowing the choice of bed height to suit the application
Features and Benefits
Flow adapters are available for each of the column diameters to allow optimal bed heights for each application, as are thermostatic jackets for precise temperature control.
Certificate of Analysis
Certificate of Origin