Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
UNSPSC Code:
41103032
Manufacturer/tradename:
Diversified Biotech DTCR-6200-Y
Material:
yellow , polypropylene
L × w:
1.28 in. × 0.50 in. × 3/8 in., spot combo
Packaging:
roll of 1000 ea (500 tag, 500 spot)
material
yellow , polypropylene
manufacturer/tradename
Diversified Biotech DTCR-6200-Y
parameter
temp. range at 70 to -196 °C
L × W
1.28 in. × 0.50 in. × 3/8 in., spot combo
packaging
roll of 1000 ea (500 tag, 500 spot)
Looking for similar products? Visit Product Comparison Guide
General description
Direct Thermal Cryo-Tags® are formatted to work with most direct thermal printers. Quickly label your samples with sharp text, clear barcodes or a combination of both. Cryogenic labels are suitable for a variety of applications.
Note: that the Dymo LabelWriters are not compatible with Direct Thermal Cryo-Tags®.
Note: that the Dymo LabelWriters are not compatible with Direct Thermal Cryo-Tags®.
Legal Information
Cryo-Tags is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service