All Photos(1)

29609-U

Hamilton® CTC LC syringes

1750N, volume 500 μL, needle size 22 ga, needle L 51 mm (2 in.)

NACRES:
NB.03

material

blunt tip stainless steel needle

needle size

22 ga

feature

needle type N (cemented needle)
point style 3

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 203349

needle L

51 mm (2 in.)

volume

500 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.

Storage Class Code

13 - Non Combustible Solids

WGK Germany

WGK 3

Flash Point F

Not applicable

Flash Point C

Not applicable

Certificate of Analysis

Certificate of Origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service