All Photos(1)
RAD406
Supelco
radiello VOC Calibration Kit (Workplace Environment)
NACRES:
NB.24
Quality Level
Featured Industry
Environmental
format
mixture
Related Categories
General description
The VOC Calibration Kit is ideal for conducting scheduled quality control runs when analyzing workplace environments. The kit consists of twelve RAD130 Cartridge Adsorbents. Three of which are blanks and nine, divided into three concentration groups, are preloaded with VOCs to simulate 8 hour exposures (480 minutes). Concentrations are described in the listed table. The values shown are indicative. Actual concentrations are certified for each lot. The composition of VOCs represents a broad range of polarity, and the spiked concentrations represent 0.5, 1.0, and 2.0 times the threshold limit values (TLV) for each compound. VOCs are spiked on the cartridges by injecting vaporized VOC standards in CS2 under nitrogen flow. The cartridges are stable for at least four months when stored at 4°C.
Legal Information
radiello is a trademark of Institi Clinici Scientifici Maugeri
Certificate of Analysis
Certificate of Origin
Passive Sampling of Atmospheric Organic Contaminants
Esteve-Turrillas, F.A., et al.
Comprehensive Sampling and Sample Preperation, 1, 201-222 (2012)
Volatile organic compounds in air
Methods for the Determination of Hazardous Substances, 1-17 (1997)
Study of the dispersion of VOCs emitted by a municipal solid waste landfill
Chiriac, R., et al.
Atmospheric Environment, 43 (11), 1926-1931 (2009)
Isabel M Morales Terrés et al.
Journal of environmental management, 91(12), 2754-2762 (2010-09-03)
This paper describes a novel methodology for evaluating the extent to which petrol stations affect their surroundings. The method is based on the fact that the ratio of the concentrations of aliphatic and aromatic hydrocarbon pollutants in the air of...
C Minoia et al.
The Science of the total environment, 192(2), 163-182 (1996-12-02)
A study using individual dosimetry to evaluate the daily inhaled dose of sixteen aromatic and aliphatic hydrocarbons in three groups of primary school children, living in three Italian towns with 50,000 inhabitants or less, (Treviglio-Lombardy; Poggibonsi-Tuscany; Valenza-Piedmont) is presented. The...
Protocols
radiello® Diffusive Air Sampling Application - Volatile Organic Compounds (VOCs) Chemically Desorbed With CS2
radiello® Diffusive Air Sampling Application - Volatile Organic Compounds (VOCs) Chemically Desorbed With CS2