Skip to Content
Merck

TA9H98DCB5EE

Targetmol Chemicals Inc.

Sucrose-epichlorohydrin copolymer

Synonym(s):

(2R,3R,4S,5S,6R)-2-(((2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol--2-(chloromethyl)oxirane (1/1)

Sign In to View Organizational & Contract Pricing.

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Inchi Code

1S/C12H22O11.C3H5ClO/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12;4-1-3-2-5-3/h4-11,13-20H,1-3H2;3H,1-2H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+;/m1./s1

Inchi Key

IACFXVUNKCXYJM-AKSHDPDZSA-N

Storage temp.

-80/-20

Country of Origin

CN

Shipping Temperature

4

Physical Form

solid

MSDS

https://www.targetmol.com/compound/sucrose-epichlorohydrin_copolymer


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service