SE
EN
Applications
Products
Services
Support
Home
Search Results
DMG-PEG 2000
(1)
DMG-PEG 2000
Empirical Formula (Hill Notation):
C
122
H
242
O
50
CAS Number:
1397695-86-1
Molecular Weight:
2509.20
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
880151P
DMG-PEG 2000, Avanti Polar Lipids
Expand
View Pricing