  • Search Results
  • Hemodialyzer mass transfer-area coefficients for urea increase at high dialysate flow rates
Hemodialyzer mass transfer-area coefficients for urea increase at high dialysate flow rates
Kidney International (1997)
Leypoldt JK, et al.