- Home
- Search Results
- Hepatic targeting and biodistribution of human fetal liver stem/progenitor cells and adult hepatocytes in mice
Hepatic targeting and biodistribution of human fetal liver stem/progenitor cells and adult hepatocytes in mice
Hepatology (2009)
Cheng K, et al.