  • Search Results
  • Cell-type-specific metabolic labeling, detection and identification of nascent proteomes in vivo.
Cell-type-specific metabolic labeling, detection and identification of nascent proteomes in vivo.
Nature protocols (2019-01-06)
Beatriz Alvarez-Castelao, Christoph T Schanzenbächer, Julian D Langer, Erin M Schuman
ABSTRACT
A big challenge in proteomics is the identification of cell-type-specific proteomes in vivo. This protocol describes how to label, purify and identify cell-type-specific proteomes in living mice. To make this possible, we created a Cre-recombinase-inducible mouse line expressing a mutant methionyl-tRNA synthetase (L274G), which enables the labeling of nascent proteins with the non-canonical amino acid azidonorleucine (ANL). This amino acid can be conjugated to different affinity tags by click chemistry. After affinity purification (AP), the labeled proteins can be identified by tandem mass spectrometry (MS/MS). With this method, it is possible to identify cell-type-specific proteomes derived from living animals, which was not possible with any previously published method. The reduction in sample complexity achieved by this protocol allows for the detection of subtle changes in cell-type-specific protein content in response to environmental changes. This protocol can be completed in ~10 d (plus the time needed to generate the mouse lines, the desired labeling period and MS analysis).
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
TWEEN® 20, viscous liquid
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Urea, powder, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Triton X-100, BioXtra
Sigma-Aldrich
Tris Buffered Saline, 10 ×, solution
Sigma-Aldrich
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine, 97%
Supelco
SYPRO® Ruby Protein Gel Stain
Sigma-Aldrich
D-(+)-Maltose monohydrate, BioXtra, ≥99%
Sigma-Aldrich
1H-1,2,3-Triazole, 97%
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O
Sigma-Aldrich
ECO TWEEN® 20, viscous liquid
Sigma-Aldrich
Light Diagnostics Measles Antibody IFA Reagent, ~50 tests, included in kit #3187, from mouse