Merck
  • Electrophilic trifluoromethylation of arenes and N-heteroarenes using hypervalent iodine reagents
Electrophilic trifluoromethylation of arenes and N-heteroarenes using hypervalent iodine reagents
Journal of Fluorine Chemistry (2010)
Wiehn, M. S.; Vinogradova, E. V.; Togni, A.