  • Search Results
  • New 16S rRNA primers to uncover Bdellovibrio and like organisms diversity and abundance.
New 16S rRNA primers to uncover Bdellovibrio and like organisms diversity and abundance.
Journal of microbiological methods (2020-07-01)
Jade A Ezzedine, Cécile Chardon, Stéphan Jacquet
ABSTRACT
Appropriate use and specific primers are important in assessing the diversity and abundance of microbial groups of interest. Bdellovibrio and like organisms (BALOs), that refer to obligate Gram-negative bacterial predators of other Gram-negative bacteria, evolved in terms of taxonomy and classification over the past two decades. Hence, some former primers have become inadequate while others are yet to be designed, for both PCR (especially with the advent of NGS) and qPCR approaches. Thus, to study BALOs' abundance and diversity in a variety of aquatic ecosystems, we designed in silico specific primer sets for each BALO genera and tested them in vitro on a variety of cultures and environmental samples. Also, we performed Sanger and Nano Miseq sequencing to reveal the exact degree of specificity of the most promising primers set. Here we report our success in designing specific primers for some BALOs genera, i.e. Bdellovibrio (PCR), Bacteriovorax (qPCR), Peredibacter (PCR).
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O