Merck
  • Synthesis, infrared spectra and thermal studies of Zn(II), Cd(II) and Hg(II) complexes with 2-aminobenzaldehyde phenylhydrazone "nitrin" ligand.
Synthesis, infrared spectra and thermal studies of Zn(II), Cd(II) and Hg(II) complexes with 2-aminobenzaldehyde phenylhydrazone "nitrin" ligand.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2007-09-04)
Moamen S Refat, Asmaa A Ibrahim
ABSTRACT
The, nitrin, 2-aminobenzaldehyde phenylhydrazone (2ABPH) was synthesis by refluxing 2-nitrobenzaldehyde with phenylhydrazine in ethanolic solvent. Three transition metal (II) complexes of 2ABPH have been prepared. Elemental analysis, molar conductivity, IR, UV, 1H NMR, and mass spectra, as well as TG/DTG have been used to characterize these complexes. The complexes have the general formula [M(2ABPH)2]Cl2.nH2O, where M=Zn, Cd, and Hg and n=4, 2 and 0 for Zn(II), Cd(II) and Hg(II), respectively. The ligand and its complexes have been studied for their possible biological activity including antibacterial and antifungal activity.
Sigma-Aldrich
2-Aminobenzaldehyde, ≥98%
Pricing and availability is not currently available.