Merck
  • Human gamma gamma-enolase: two-site immunoradiometric assay with a single monoclonal antibody.
Human gamma gamma-enolase: two-site immunoradiometric assay with a single monoclonal antibody.
Journal of neurochemistry (1987-01-01)
B S Soler-Federsppiel, P Cras, J Gheuens, D Andries, A Lowenthal
ABSTRACT
A monoclonal antibody (mAb), termed BBS/NC/VI-H14 (H14), that reacts with the human enzyme gamma gamma-enolase was prepared. It was directed against the gamma-subunit and did not cross-react with the alpha- or beta-subunit. The mAb H14 can be used for quantitative determination of gamma gamma-enolase in a two-site immunoradiometric assay (two-site IRMA). It is also suitable for immunostaining formalin-fixed tissues. The specific identification of gamma gamma-enolase provided by the two-site IRMA with H14 is discussed in relation to the cellular distribution of this protein.