  • Search Results
  • Adaptive and innate immune responses in a rat orthotopic lung transplant model of chronic lung allograft dysfunction.
Adaptive and innate immune responses in a rat orthotopic lung transplant model of chronic lung allograft dysfunction.
Transplant international : official journal of the European Society for Organ Transplantation (2014-09-03)
Alena Evers, Srebrena Atanasova, Gabriele Fuchs-Moll, Kathrin Petri, Sigrid Wilker, Anna Zakrzewicz, Markus Hirschburger, Winfried Padberg, Veronika Grau
ABSTRACT
Acute rejection and respiratory infections are major risk factors for chronic lung allograft dysfunction (CLAD) after lung transplantation. To shed light on the enigmatic etiology of CLAD, we test the following hypotheses using a new experimental model: (i) Alloimmune-independent pulmonary inflammation reactivates alloimmunity. (ii) Alloimmunity enhances the susceptibility of the graft toward pathogen-associated molecular patterns. Pulmonary Fischer 344 to Lewis rat allografts were treated with lipopolysaccharide (LPS), which consistently results in lesions typical for CLAD. Grafts, local lymph nodes, and spleens were harvested before (day 28) and after LPS application (days 29, 33, and 40) for real-time RT-PCR and immunohistochemistry. Mixed lymphocyte reactions were performed on day 33. Four weeks after transplantation, lung allografts displayed mononuclear infiltrates compatible with acute rejection and overexpressed most components of the toll-like receptor system. Allografts but not secondary lymphoid organs expressed increased levels of Th1-type transcription factors and cytokines. LPS induced macrophage infiltration as well as mRNA expression of pro-inflammatory cytokines and effector molecules of innate immunity. Unexpectedly, T-cell reactivity was not enhanced by LPS. We conclude that prevention of CLAD might be accomplished by local suppression of Th1 cells in stable grafts and by controlling innate immunity during alloimmune-independent pulmonary inflammation.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥970 μg/mg (USP XXIV)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material