  • Search Results
  • Analysis of expressed genes of the bacterium 'Candidatus phytoplasma Mali' highlights key features of virulence and metabolism.
Analysis of expressed genes of the bacterium 'Candidatus phytoplasma Mali' highlights key features of virulence and metabolism.
PloS one (2014-04-15)
Christin Siewert, Toni Luge, Bojan Duduk, Erich Seemüller, Carmen Büttner, Sascha Sauer, Michael Kube
ABSTRACT
'Candidatus Phytoplasma mali' is a phytopathogenic bacterium of the family Acholeplasmataceae assigned to the class Mollicutes. This causative agent of the apple proliferation colonizes in Malus domestica the sieve tubes of the plant phloem resulting in a range of symptoms such as witches'--broom formation, reduced vigor and affecting size and quality of the crop. The disease is responsible for strong economical losses in Europe. Although the genome sequence of the pathogen is available, there is only limited information on expression of selected genes and metabolic key features that have not been examined on the transcriptomic or proteomic level so far. This situation is similar to many other phytoplasmas. In the work presented here, RNA-Seq and mass spectrometry shotgun techniques were applied on tissue samples from Nicotiana occidentalis infected by 'Ca. P. mali' strain AT providing insights into transcriptome and proteome of the pathogen. Data analysis highlights expression of 208 genes including 14 proteins located in the terminal inverted repeats of the linear chromosome. Beside a high portion of house keeping genes, the recently discussed chaperone GroES/GroEL is expressed. Furthermore, gene expression involved in formation of a type IVB and of the Sec-dependent secretion system was identified as well as the highly expressed putative pathogenicity-related SAP11-like effector protein. Metabolism of phytoplasmas depends on the uptake of spermidine/putescine, amino acids, co-factors, carbohydrates and in particular malate/citrate. The expression of these transporters was confirmed and the analysis of the carbohydrate cycle supports the suggested alternative energy-providing pathway for phytoplasmas releasing acetate and providing ATP. The phylogenetic analyses of malate dehydrogenase and acetate kinase in phytoplasmas show a closer relatedness to the Firmicutes in comparison to Mycoplasma species indicating an early divergence of the Acholeplasmataceae from the Mollicutes.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
USP
Sodium acetate, United States Pharmacopeia (USP) Reference Standard