Merck
Novel high-throughput deoxyribonuclease 1 assay.
Journal of biomolecular screening (2014-10-19)
Dae Song Jang, Narsimha R Penthala, Eugene O Apostolov, Xiaoying Wang, Tariq Fahmi, Peter A Crooks, Alexei G Basnakian
ABSTRACT
Deoxyribonuclease I (DNase I), the most active and abundant apoptotic endonuclease in mammals, is known to mediate toxic, hypoxic, and radiation injuries to the cell. Neither inhibitors of DNase I nor high-throughput methods for screening of high-volume chemical libraries in search of DNase I inhibitors are, however, available. To overcome this problem, we developed a high-throughput DNase I assay. The assay is optimized for a 96-well plate format and based on the increase of fluorescence intensity when fluorophore-labeled oligonucleotide is degraded by the DNase. The assay is highly sensitive to DNase I compared to other endonucleases, reliable (Z' ≥ 0.5), and operationally simple, and it has low operator, intraassay, and interassay variability. The assay was used to screen a chemical library, and several potential DNase I inhibitors were identified. After comparison, 2 hit compounds were selected and shown to protect against cisplatin-induced kidney cell death in vitro. This assay will be suitable for identifying inhibitors of DNase I and, potentially, other endonucleases.
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%