  • Search Results
  • The histone demethylase KDM4B interacts with MyoD to regulate myogenic differentiation in C2C12 myoblast cells.
The histone demethylase KDM4B interacts with MyoD to regulate myogenic differentiation in C2C12 myoblast cells.
Biochemical and biophysical research communications (2014-12-24)
Jang Hyun Choi, Young Joon Song, Hansol Lee
ABSTRACT
Enzymes that mediate posttranslational modifications of histone and nonhistone proteins have been implicated in regulation of skeletal muscle differentiation. However, functions of histone demethylases that could counter the actions of H3-K9 specific histone methyltransferases remain still obscure. Here we present evidences that KDM4B histone demethylase regulates expression of myogenic regulators such as MyoD and thereby controls myogenic differentiation of C2C12 myoblast cells. We demonstrate that expression of KDM4B gradually increases during myogenic differentiation and depletion of KDM4B using shRNA results in inhibition of differentiation in C2C12 myoblast cells, which is correlated with decreased expression of MyoD and myogenin. In addition, we find that KDM4B shRNA represses expression of MyoD promoter-driven luciferase reporter and exogenous expression of MyoD rescues myogenic potential in KDM4B-depleted myoblast cells. We further show that KDM4B interacts with MyoD, binds to MyoD and myogenin promoters in vivo, and finally, is involved in demethylation of tri-methylated H3-K9 on promoters of MyoD and myogenin. Taken together, our data suggest that KDM4B plays key roles in myogenic differentiation of C2C12 cells, presumably by its function as a H3-K9 specific histone demethylase.
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Supelco
Sodium acetate solution, NMR reference standard, 50 mM in D2O (99.9 atom % D), water 1 %, NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%