  • Search Results
  • Reactive species and oxidative stress in optic nerve vulnerable to secondary degeneration.
Reactive species and oxidative stress in optic nerve vulnerable to secondary degeneration.
Experimental neurology (2014-06-17)
Ryan L O'Hare Doig, Carole A Bartlett, Ghassan J Maghzal, Magdalena Lam, Michael Archer, Roland Stocker, Melinda Fitzgerald
ABSTRACT
Secondary degeneration contributes substantially to structural and functional deficits following traumatic injury to the CNS. While it has been proposed that oxidative stress is a feature of secondary degeneration, contributing reactive species and resultant oxidized products have not been clearly identified in vivo. The study is designed to identify contributors to, and consequences of, oxidative stress in a white matter tract vulnerable to secondary degeneration. Partial dorsal transection of the optic nerve (ON) was used to model secondary degeneration in ventral nerve unaffected by the primary injury. Reactive species were assessed using fluorescent labelling and liquid chromatography/tandem mass spectroscopy (LC/MS/MS). Antioxidant enzymes and oxidized products were semi-quantified immunohistochemically. Mitophagy was assessed by electron microscopy. Fluorescent indicators of reactive oxygen and/or nitrogen species increased at 1, 3 and 7days after injury, in ventral ON. LC/MS/MS confirmed increases in reactive species linked to infiltrating microglia/macrophages in dorsal ON. Similarly, immunoreactivity for glutathione peroxidase and haem oxygenase-1 increased in ventral ON at 3 and 7days after injury, respectively. Despite increased antioxidant immunoreactivity, DNA oxidation was evident from 1day, lipid oxidation at 3days, and protein nitration at 7days after injury. Nitrosative and oxidative damage was particularly evident in CC1-positive oligodendrocytes, at times after injury at which structural abnormalities of the Node of Ranvier/paranode complex have been reported. The incidence of mitochondrial autophagic profiles was also significantly increased from 3days. Despite modest increases in antioxidant enzymes, increased reactive species are accompanied by oxidative and nitrosative damage to DNA, lipid and protein, associated with increasing abnormal mitochondria, which together may contribute to the deficits of secondary degeneration.
Sigma-Aldrich
3-Nitro-L-tyrosine, crystalline
Sigma-Aldrich
Dihydroethidium, ≥95%
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard