  • Search Results
  • Antidepressant-like effect of bright light is potentiated by L-serine administration in a mouse model of seasonal affective disorder.
Antidepressant-like effect of bright light is potentiated by L-serine administration in a mouse model of seasonal affective disorder.
Brain research bulletin (2015-09-06)
Misato Kawai, Ryosei Goda, Tsuyoshi Otsuka, Ayaka Iwamoto, Nobuo Uotsu, Mitsuhiro Furuse, Shinobu Yasuo
ABSTRACT
Bright light therapy is used as the primary treatment for seasonal affective disorder; however, the mechanisms underlying its antidepressant effect are not fully understood. Previously, we found that C57BL/6J mice exhibit increased depression-like behavior during a short-day condition (SD) and have lowered brain serotonin (5-HT) content. This study analyzed the effect of bright light on depression-like behaviors and the brain serotonergic system using the C57BL/6J mice. In the mice maintained under SD, bright light treatment (1000 lx, daily 1 h exposure) for 1 week reduced immobility time in the forced swimming test and increased intake of saccharin solution in a saccharin intake test. However, the light treatment did not modify 5-HT content and selective 5-HT uptake in the amygdala, or temporal patterns of core body temperature and wheel-running activity throughout a day. In the next experiment, we attempted to enhance the effect of bright light by using L-serine, a precursor of D-serine that acts as an N-methyl-D-aspartic acid receptor coagonist. Daily subcutaneous injection of L-serine for 2 weeks prior to the bright light strongly reduced the immobility time in the forced swimming test, suggesting a synergistic effect of light and L-serine. Furthermore, bright light increased the total number of 5-HT-immunoreactive cells and cells that had colocalized 5-HT and c-Fos immunosignals in several subregions of the raphe nuclei. These effects were potentiated by prior injection of L-serine. These data suggest that the bright light may elicit an antidepressant-like effect via enhanced 5-HT signals in the brain and L-serine can enhance these effects.
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%