  • Search Results
  • Matrix metalloproteinase 13: a potential intermediate between low expression of microRNA-125b and increasing metastatic potential of non-small cell lung cancer.
Matrix metalloproteinase 13: a potential intermediate between low expression of microRNA-125b and increasing metastatic potential of non-small cell lung cancer.
Cancer genetics (2015-03-17)
Xiaozhou Yu, Feng Wei, Jinpu Yu, Hua Zhao, Lei Jia, Yingnan Ye, Ruijuan Du, Xiubao Ren, Hui Li
ABSTRACT
Recent findings have suggested that microRNAs may be involved in the regulation of metastasis in malignant cancers such as non-small cell lung cancer (NSCLC). This study aimed to determine the relationship between expression of miR-125b and matrix metalloproteinase 13 (encoded by MMP-13) and the metastatic potential of cancer cells in NSCLC. Expression levels of miR-125b transcripts and MMP-13 proteins were analyzed by quantitative real-time PCR and immunohistochemistry, respectively, in tumor tissues and adjacent nontumor tissues from 42 patients with NSCLC. The interaction between miR-125b and MMP-13 expression and the associations between miR-125b and clinicopathologic data were analyzed. MiR-125 b expression levels were decreased in NSCLC tumor tissue samples, which correlated with an increased incidence of lymph node metastases, increased pathologic stage, increased MMP-13 expression levels, and decreased early progression-free survival. Additionally, we have demonstrated that increased levels of miR-125b can directly downregulate MMP-13 protein expression and inhibit the invasive capabilities of cancer cells. Expression levels of miR-125b were negatively correlated with metastatic potential of NSCLC tumors, which may function through regulation of MMP-13.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O