  • Search Results
  • Improved revascularization of islet grafts using an angiogenic monocyte subpopulation derived from spheroid culture of bone marrow mononuclear cells.
Improved revascularization of islet grafts using an angiogenic monocyte subpopulation derived from spheroid culture of bone marrow mononuclear cells.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2015-04-14)
B J Oh, S-M Jin, J-M Choi, S-H Oh, W Shim, M-S Lee, M-K Lee, J H Kim
ABSTRACT
The spheroid culture method is an effective strategy for ex vivo expansion of an autologous therapeutic cell population. We investigated if cotransplantation of bone marrow-derived spheroids (BM-spheroid) formed using 3D culture of BM-derived mononuclear cells (BM-MNCs) could improve the posttransplant outcome of islet grafts using a mouse syngeneic marginal mass renal subcapsular islet transplantation model. Using green fluorescent protein transgenic (GFP-Tg) mice, the role of the BM-spheroids and the contribution of vessels derived from donors and recipients in grafted areas were assessed by immunohistochemistry. Compared to fresh BM-MNCs and nonspheroid remnant cells (BM-nonspheroid), the BM-spheroids, mainly composed of CXCR4(+) CD14(+) myeloid cells, showed higher angiogenic capacity, such as in vitro self-formed vessel structures; increased expression of angiogenic and chemoattractive factors; and incorporation into new vessel formation in basement membrane matrix plugs. BM-spheroid cotransplantation with islets improved the posttransplant outcomes in terms of glucose tolerance, serum insulin level, and diabetes reversal rate when compared with cotransplantation of BM-nonspheroids. Immunohistochemistry revealed that cotransplantation of the BM-spheroids increased vessel density, area of grafted endocrine and non-endocrine tissue, and β cell proliferation. In conclusion, cotransplantation of islets and BM-spheroids improved islet function through facilitation of revascularization and an increase in cell proliferation and islet cell mass.
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder
Sigma-Aldrich
Glucagon, synthetic, powder, suitable for cell culture
Sigma-Aldrich
Glucagon, ≥95% (HPLC), powder, synthetic