Merck
Wheat milling by-products and their impact on bread making.
Food chemistry (2015-05-16)
Sami Hemdane, Sofie Leys, Pieter J Jacobs, Emmie Dornez, Jan A Delcour, Christophe M Courtin
ABSTRACT
This study investigates the relationship between the properties of dietary fiber (DF) rich wheat milling by-products and their impact on bread making. From coarse bran over coarse and fine weatings to low grade flour, the content of starch and lipids increased, while that of DF and ash decreased. Enzyme activity levels differed strongly and were not related to other by-product properties. Average particle size of the by-products was positively correlated with DF and ash contents and their hydration properties. When meals from flour and by-products were composed on the same overall starch level to compensate for differences in endosperm contamination in the by-products, bread specific volume was more strongly depressed with fine weatings and low grade flour than with coarse bran and weatings. This suggests that the properties of the former were intrinsically more detrimental to bread making than those of the latter.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, meets USP testing specifications
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FCC, FG
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%