  • Search Results
  • Anti-Inflammatory and Anti-Thrombotic Effects of the Fungal Metabolite Galiellalactone in Apolipoprotein E-Deficient Mice.
Anti-Inflammatory and Anti-Thrombotic Effects of the Fungal Metabolite Galiellalactone in Apolipoprotein E-Deficient Mice.
PloS one (2015-06-16)
Franziska Bollmann, Sven Jäckel, Lisa Schmidtke, Katharina Schrick, Christoph Reinhardt, Kerstin Jurk, Zhixiong Wu, Ning Xia, Huige Li, Gerhard Erkel, Ulrich Walter, Hartmut Kleinert, Andrea Pautz
ABSTRACT
Patients suffering from chronic inflammatory diseases have an increased mortality risk resulting from cardiovascular disorders due to enhanced atherosclerotic and thrombotic events. Until now, it is not completely understood in which way an abnormal expression of pro-inflammatory mediators contributes to this elevated cardiovascular risk, but there is a need for new drugs that on the one hand suppress the expression of pro-inflammatory mediators and on the other hand inhibit arterial platelet adhesion. Thus, we analyzed the anti-inflammatory and anti-thrombotic capacity of the fungal metabolite Galiellalactone in atherosclerosis-prone apolipoprotein E-deficient mice. Treatment of the mice with Galiellalactone lowered the inflammatory expression profile and improved blood clotting times, as well as platelet adhesion to the injured common carotid artery. The results indicate that administration of Galiellalactone is able to reduce the extent of inflammation and arterial platelet adhesion in this mouse model.
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)