  • Search Results
  • The nod-like receptor, Nlrp12, plays an anti-inflammatory role in experimental autoimmune encephalomyelitis.
The nod-like receptor, Nlrp12, plays an anti-inflammatory role in experimental autoimmune encephalomyelitis.
Journal of neuroinflammation (2015-11-02)
Marjan Gharagozloo, Tara M Mahvelati, Emilie Imbeault, Pavel Gris, Echarki Zerif, Diwakar Bobbala, Subburaj Ilangumaran, Abdelaziz Amrani, Denis Gris
ABSTRACT
Multiple sclerosis (MS) is an organ-specific autoimmune disease resulting in demyelinating plaques throughout the central nervous system. In MS, the exact role of microglia remains unknown. On one hand, they can present antigens, skew T cell responses, and upregulate the expression of pro-inflammatory molecules. On the other hand, microglia may express anti-inflammatory molecules and inhibit inflammation. Microglia express a wide variety of immune receptors such as nod-like receptors (NLRs). NLRs are intracellular receptors capable of regulating both innate and adaptive immune responses. Among NLRs, Nlrp12 is largely expressed in cells of myeloid origins. It plays a role in immune inflammatory responses by negatively regulating the nuclear factor-kappa B (NF-κB) pathway. Thus, we hypothesize that Nlrp12 suppresses inflammation and ameliorates the course of MS. We used experimental autoimmune encephalomyelitis (EAE), a well-characterized mouse model of MS. EAE was induced in wild-type (WT) and Nlrp12 (-/-) mice with myelin oligodendrocyte glycoprotein (MOG):complete Freud's adjuvant (CFA). The spinal cords of healthy and immunized mice were extracted for immunofluorescence and pro-inflammatory gene analysis. Primary murine cortical microglia cell cultures of WT and Nlrp12 (-/-) were prepared with cortices of 1-day-old pups. The cells were stimulated with lipopolysaccharide (LPS) and analyzed for the expression of pro-inflammatory genes as well as pro-inflammatory molecule secretions. Over the course of 9 weeks, the Nlrp12 (-/-) mice demonstrated increased severity in the disease state, where they developed the disease earlier and reached significantly higher clinical scores compared to the WT mice. The spinal cords of immunized WT mice relative to healthy WT mice revealed a significant increase in Nlrp12 messenger ribonucleic acid (mRNA) expression at 1, 3, and 5 weeks post injection. A significant increase in the expression of pro-inflammatory genes Ccr5, Cox2, and IL-1β was found in the spinal cords of the Nlrp12 (-/-) mice relative to the WT mice (P < 0.05). A significant increase in the level of gliosis was observed in the spinal cords of the Nlrp12 (-/-) mice compared to the WT mice after 9 weeks of disease (P < 0.05). Primary Nlrp12 (-/-) microglia cells demonstrated a significant increase in inducible nitric oxide synthase (iNOS) expression (P < 0.05) and secreted significantly (P < 0.05) more tumor necrosis factor alpha (TNFα), interleukin-6 (IL-6), and nitric oxide (NO). Nlrp12 plays a protective role by suppressing inflammation during the development of EAE. The absence of Nlrp12 results in an increased inflammatory response.
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
o-Xylene, reagent grade, ≥98.0%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, for molecular biology
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)