  • Search Results
  • Evaluation of the protective efficacy of four novel identified membrane associated proteins of Streptococcus suis serotype 2.
Evaluation of the protective efficacy of four novel identified membrane associated proteins of Streptococcus suis serotype 2.
Vaccine (2015-03-31)
Yang Zhou, Yan Wang, Limei Deng, Chengkun Zheng, Fangyan Yuan, Huanchun Chen, Weicheng Bei, Jinquan Li
ABSTRACT
Streptococcus suis serotype 2 (S. suis 2) is an important zoonotic pathogen that can also cause epidemics of life-threatening infections in humans. Surface proteins of pathogens play a critical role in the interaction with host system or environment, as they take part in processes like virulence, cytotoxicity, adhesion, signaling or transport, etc. Thus, surface proteins identified by the screening of immunoproteomic techniques are promising vaccine candidates or diagnostic markers. In this study, four membrane associated proteins (MAP) identified by immunoproteomic method were cloned and expressed as recombinant proteins with his-tag. Screening for vaccine candidates were firstly performed by protection assay in vivo and immunization with Sbp markedly protected mice against systemic S. suis 2 infection. The immune responses and protective of Sbp were further evaluated. The results showed that Sbp could elicit a strong humoral antibody response and protect mice from lethal challenge with S. suis 2. The antiserum against Sbp could efficiently impede survival of bacterial in whole blood killing assay and conferred significant protection against S. suis 2 infection in passive immunization assays. The findings indicate that Sbp may serve as an important factor in the pathogenesis of S. suis 2 and would be a promising subunit vaccine candidate.
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O