  • Search Results
  • Anticancer activity of CopA3 dimer peptide in human gastric cancer cells.
Anticancer activity of CopA3 dimer peptide in human gastric cancer cells.
BMB reports (2014-07-23)
Joon Ha Lee, In-Woo Kim, Sang-Hee Kim, Eun-Young Yun, Sung-Hee Nam, Mi-Young Ahn, Dong-Chul Kang, Jae Sam Hwang
ABSTRACT
CopA3 is a homodimeric α-helical peptide derived from coprisin which is a defensin-like antimicrobial peptide that was identified from the dung beetle, Copris tripartitus. CopA3 has been reported to have anticancer activity against leukemia cancer cells. In the present study, we investigated the anticancer activity of CopA3 in human gastric cancer cells. CopA3 reduced cell viability and it was cytotoxic to gastric cancer cells in the MTS and LDH release assay, respectively. CopA3 was shown to induce necrotic cell death of the gastric cancer cells by flow cytometric analysis and acridine orange/ethidium bromide staining. CopA3-induced cell death was mediated by specific interactions with phosphatidylserine, a membrane component of cancer cells. Taken together, these data indicated that CopA3 mainly caused necrosis of gastric cancer cells, probably through interactions with phosphatidylserine, which suggests the potential utility of CopA3 as a cancer therapeutic.
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Citric acid trisodium salt, anhydrous, ≥98% (GC)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Agarose, Type I, low EEO