- NF-kappaB-dependent control of HIV-1 transcription by the second coding exon of Tat in T cells.
NF-kappaB-dependent control of HIV-1 transcription by the second coding exon of Tat in T cells.
HIV-1 two-exon transactivator protein (Tat) is a 101-aa protein. We investigated the possible contribution of the extreme C terminus of HIV-1 Tat to maximize nuclear transcription factor NF-kappaB activation, long terminal repeat (LTR) transactivation, and viral replication in T cells. C-terminal deletion and substitution mutants made with the infectious clone HIV-89.6 were assayed for their ability to transactivate NF-kappaB-secreted alkaline phosphatase and HIV-1 LTR-luciferase reporter constructs for low concentrations of Tat. A mutant infectious clone of HIV-89.6 engineered by introducing a stop codon at aa 72 in the Tat open-reading frame (HIVDeltatatexon2) replicated at a significantly lower rate than the wild-type HIV-89.6 in phytohemagglutinin-A/IL-2-stimulated primary peripheral blood lymphocytes. Altogether, our results suggest a critical role for the glutamic acids at positions 92, 94, and 96 or lysines at positions 88, 89, and 90, present in the second encoding Tat exon in activating NF-kappaB, transactivating the HIV-1 LTR and enhancing HIV-1 replication in T cells.