Merck
HomeNHC-Based Pd Catalysts and Ligands for C–C Bond Formation
NHC-Based Pd Catalysts and Ligands for C–C Bond Formation
In collaboration with Umicore,1 we are pleased to offer a series of robust Pd(II) and Pd(0) complexes employed as efficient catalysts in C–C bond forming reactions. The high performance Pd catalysts reviewed below can rapidly couple alkyl and aryl chlorides with organoboron compounds on large scale (100 g-100 t/a).2 The high TONs, mild reaction conditions, and economic viability/availability of aryl chlorides, make this methodology attractive to industrial scale applications. Catalysts 11 and 12 (Scheme 14) should exhibit superior activity in C–C coupling reactions, because they are formally Pd(0) and are rare examples of well-characterized monocarbene palladium precursors to 12‑electron complexes. Indeed, the Umicore NHC-Pd system performs Suzuki and Kumada couplings as well as a-arylation reactions at mild temperatures.
nhc-based-pd-catalysts
Scheme 14. (660388) (660272)
In the latter case, (NHC)Pd(allyl)Cl (13),2 a reactive, formally 16‑electron complex, mediates the a-arylation of an array of aryl ketones (Scheme 15).3 The air-stable catalyst, short reaction times, and high conversions prove the usefulness of this NHC technology over previous Pd systems. This system can be optimized by utilizing excess aryl halides which, in turn, increases the reaction rates and ensures high product yields in as little as 15 min. Reactivity of both alkyl–alkyl and alkyl–aryl ketones was studied in the early NHC-Pd article from Nolan and co-workers.
nhc-based-pd-catalysts1
Scheme 15.
[Pd(IMes)(NQ)]2 catalyst 12 demonstrated high reactivity and selectivity in sp3–sp2 Kumada couplings.4 The generality of this methodology extends to both electron-rich and electron-poor aryl magnesium reagents. Furthermore, a broad spectrum of functionalized alkyl chlorides was employed to afford complex organic building blocks (Scheme 16). The high product yields at room temperature validates the robustness of this catalytic system versus well-known Pd-phosphine catalysts Pd(PPh3)4 and Pd2(dba3) as a function of reaction conditions.
nhc-based-pd-catalysts2
Scheme 16.
The related [Pd(IPr)(NQ)]2 catalyst 11 exhibited impressive activity in the Suzuki–Miyaura coupling of aryl chlorides with phenyl boronic acid (Scheme 17). At 50 °C, the high-yielding (88%) reaction was complete in one hour at a catalyst loading of 0.5 mol %.5 Interestingly, Pd(0) catalyst 11 produced lower yields of coupled biaryl product at room temperature, whereas analogous catalyst 12 gave 86% yields of 4-Me-biphenyl at both room temperature and 50 °C under identical loadings conditions. Presumably, catalyst 11 needs additional energy to climb over the activation barrier and enter the catalytic cycle as a naked Pd-NHC species. It should be noted that the reactivity of [Pd(IPr)(NQ)]2 was also shown to be high in the coupling of sterically encumbered 2,6-diphenyl chloride and 1‑naphthalene boronic acid (Scheme 18).
nhc-based-pd-catalysts3
Scheme 17.
nhc-based-pd-catalysts4
Scheme 18.
Beller and co-workers succeeded in establishing a reactivity profile for NHC-Pd naphtholquinone catalysts in Heck reactions (Table 4).6 The outstanding capacity of this system is illustrated in Scheme 19, wherein good stillbene yields were obtained at 140 °C in an ionic liquid media. The low catalyst loading (0.5 mol %), cheap aryl chloride reagents, and a stabilized ionic liquid environment all contribute to the potential advancement of this chemistry to the industrial fine chemical arena.
Table 4.
nhc-based-pd-catalysts5
Scheme 19.
NHC Ligands
It is well known that the activity of Pd catalysts can be modified by the introduction of sterically encumbered groups approximate to the metal center. We offer two NHC ligands that contain bulky, dissimilar moieties that will impart greater catalyst design flexibility. These asymmetric ligands expand our commercial line of NHC ligands, granting ready access to a range of highly active catalysts in various important organic transformations when combined with metal precursors.
Materials
Loading
References
1.
Viciu MS, Germaneau RF, Navarro-Fernandez O, Stevens ED, Nolan SP. 2002. Activation and Reactivity of (NHC)Pd(allyl)Cl (NHC = N-Heterocyclic Carbene) Complexes in Cross-Coupling Reactions. Organometallics. 21(25):5470-5472. http://dx.doi.org/10.1021/om020804i
2.
Viciu MS, Germaneau RF, Nolan SP. 2002. Well-Defined, Air-Stable (NHC)Pd(Allyl)Cl (NHC = N-Heterocyclic Carbene) Catalysts for the Arylation of Ketones. Org. Lett.. 4(23):4053-4056. http://dx.doi.org/10.1021/ol026745m
3.
Frisch AC, Beller M. 2005. Catalysts for Cross-Coupling Reactions with Non-activated Alkyl Halides. Angew. Chem. Int. Ed.. 44(5):674-688. http://dx.doi.org/10.1002/anie.200461432
4.
Navarro O, Oonishi Y, Kelly RA, Stevens ED, Briel O, Nolan SP. 2004. General and efficient methodology for the Suzuki?Miyaura reaction in technical grade 2-propanol. Journal of Organometallic Chemistry. 689(23):3722-3727. http://dx.doi.org/10.1016/j.jorganchem.2004.04.001
5.
Selvakumar K, Zapf A, Beller M. 2002. New Palladium Carbene Catalysts for the Heck Reaction of Aryl Chlorides in Ionic Liquids. Org. Lett.. 4(18):3031-3033. http://dx.doi.org/10.1021/ol020103h