Merck
  • 685941
All Photos(2)

685941

Sigma-Aldrich

Titanium-manganese alloy

greener alternative

hydrogen-storage grade

Synonym(s):
TiMn2 type, Laves phase
Linear Formula:
TiMn2
CAS Number:
PubChem Substance ID:
NACRES:
NA.23

grade

hydrogen-storage grade

form

granular

composition

Mn, ~51%
Ti, ~29%
V, ~14%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

greener alternative category

SMILES string

[Ti].[Mn]

InChI

1S/Mn.Ti

InChI key

MECMQNITHCOSAF-UHFFFAOYSA-N

General description

Titanium-manganese alloy (TiMn2) is a titanium alloy, with properties that include high specific strength, good corrosion resistance, and biocompatibility. In this, manganese is used as a beta-stabilizer and a promising material for the alloying of titanium.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

TiMn2 alloy can be used for a variety of applications such as aerospace and hydrogen storage. Due to its biocompatibility, it finds potential usage as a biomaterial for biomedical applications.

Packaging

10 g in glass bottle

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

More Documents

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service