Merck
  • C3605
All Photos(1)

C3605

Sigma-Aldrich

Coelenterazine ip

solid

Synonym(s):
CLZN-ip
NACRES:
NA.32

form

solid

Quality Level

storage temp.

−20°C

Compare Similar Items

View Full Comparison

1 of 4

This Item
C2855SMB00232PHR1521
Sigma-Aldrich

Sigma-Aldrich

C3605

Coelenterazine ip

Sigma-Aldrich

Sigma-Aldrich

C2855

Coelenterazine cp

Clusiacitran A ≥95% (LC/MS-ELSD)

Sigma-Aldrich

SMB00232

Clusiacitran A

Supelco

Supelco

PHR1521

Ceftiofur Sodium

form

solid

form

solid

form

solid

form

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-10 to -25°C

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

300

Biochem/physiol Actions

Almost 50 times higher luminescence intensity than native coelenteraztine (Prod. No. C2230) but with a slower response time.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service