SG
EN
Applications
Products
Services
Documents
Support
Home
Search Results
Sucrose
(16)
Sucrose
Synonyms:
α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar
Empirical Formula (Hill Notation):
C
12
H
22
O
11
CAS Number:
57-50-1
Molecular Weight:
342.30
EC Number:
200-334-9
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Showing 1-16 of 16
Page 1 of 1
Page 1 of 1
Compare
Product Number
Product Description
Pricing
S0389
Sucrose,
for molecular biology, ≥99.5% (GC)
Expand
View Pricing
S9378
Sucrose,
≥99.5% (GC)
Expand
View Pricing
PHR1001
Sucrose,
Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
S7903
Sucrose,
BioXtra, ≥99.5% (GC)
Expand
View Pricing
84097
Sucrose,
BioUltra, for molecular biology, ≥99.5% (HPLC)
Expand
View Pricing
S5016
Sucrose,
ACS reagent
Expand
View Pricing
S8501
Sucrose,
≥99.5%
Expand
View Pricing
1623637
Sucrose,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
84100
Sucrose,
suitable for microbiology, ACS reagent, ≥99.0%
Expand
View Pricing
16104
Sucrose,
puriss., meets analytical specification of Ph. Eur., BP, NF
Expand
View Pricing
S1888
Sucrose,
BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.5% (GC)
Expand
View Pricing
S5391
Sucrose,
Grade II, suitable for plant cell culture
Expand
View Pricing
S5390
Sucrose,
Grade I, suitable for plant cell culture
Expand
View Pricing
S3929
Sucrose,
meets USP testing specifications
Expand
View Pricing
S1174
Sucrose,
analytical standard, for enzymatic assay kit SCA20
Expand
View Pricing
S1600000
Sucrose,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
Page 1 of 1
Page 1 of 1