SG
EN
Applications
Products
Services
Support
Home
Search Results
5-Azido-2-nitrobenzoic acid N-hydroxysuccinimide ester
(1)
5-Azido-2-nitrobenzoic acid N-hydroxysuccinimide ester
Synonyms:
N-(5-Azido-2-nitrobenzoyloxy)succinimide, N-Succinimidyl 5-azido-2-nitrobenzoate
Empirical Formula (Hill Notation):
C
11
H
7
N
5
O
6
CAS Number:
60117-35-3
Molecular Weight:
305.20
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
A3282
5-Azido-2-nitrobenzoic acid N-hydroxysuccinimide ester, ≥95%, powder
Expand
View Pricing