Merck
  • Ubiquitin Specific Protease 29 Functions as an Oncogene Promoting Tumorigenesis in Colorectal Carcinoma.
Ubiquitin Specific Protease 29 Functions as an Oncogene Promoting Tumorigenesis in Colorectal Carcinoma.
Cancers (2021-06-03)
Arun Pandian Chandrasekaran, Bharathi Suresh, Neha Sarodaya, Na-Re Ko, Seung-Jun Oh, Kye-Seong Kim, Suresh Ramakrishna
ABSTRACT
Colorectal carcinoma is the third foremost cause of cancer-related deaths and accounts for 5.8% of all deaths globally. The molecular mechanisms of colon cancer progression and metastasis control are not well studied. Ubiquitin-specific protease 29 (USP29), a deubiquitinating enzyme, is involved in the occurrence and development of wide variety of cancers. However, its clinical significance and biological roles in colorectal carcinoma (CRC) remain unexplored. In this research, we observed that the rate of USP29 overexpression was higher in colon cancer patient tissues relative to its corresponding normal tissues. CRISPR-Cas9-mediated depletion of USP29 triggered DNA double strand breaks and delayed cell-cycle progression in HCT116 cells. We also demonstrated that USP29 depletion hampers the colony formation and increases apoptosis of HCT116 cells. USP29 knockdown significantly decreased CRC cell proliferation in vitro. Depletion of USP29 in HCT116 cells substantially reduced the tumor volume of mouse xenografts. In conclusion, our study shows that elevated expression of USP29 promotes malignancy in CRC, suggesting that USP29 could be a promising target for colon cancer therapy.
Sigma-Aldrich
Anti-USP29 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution