Merck
  • Immunohistochemical detection of tartrate-resistant acid phosphatase in non-hematopoietic human tissues.
Immunohistochemical detection of tartrate-resistant acid phosphatase in non-hematopoietic human tissues.
American journal of clinical pathology (1995-10-01)
H Yaziji, A J Janckila, S C Lear, A W Martin, L T Yam
ABSTRACT
Immunohistochemical studies were done on formalin-fixed, paraffin-embedded tissues to evaluate the specificity of a newly developed monoclonal antibody (9C5) against tartrate-resistant acid phosphatase. Sections from 195 specimens were examined, which included 33 types of tissues/organs. These tissues included normal, inflammatory, and neoplastic processes. Neoplastic tissues from 14 patients with hairy cell leukemia served as positive controls. Epitope enhancement was accomplished either by microwave irradiation in citrate buffer or by boiling in water followed by trypsin digestion. Tissues were reacted with monoclonal antibody 9C5 and stained with either the avidin-biotin peroxidase method or the alkaline phosphatase anti-alkaline phosphatase method. The hairy cells of all cases of hairy cell leukemia reacted positively with 9C5. Other positively stained cells included osteoclasts, activated macrophages and giant cells. Immunohistochemical studies with 9C5, when interpreted within the context of the specificity of this antibody, are useful for the diagnosis and assessment of treatment results for hairy cell leukemia. Monoclonal antibody 9C5 also may be useful as a marker for osteoclasts and the activated macrophages and for the diagnosis of disorders involved by these cells.