Merck
  • Identification of phenolic antioxidants and bioactives of pomegranate seeds following juice extraction using HPLC-DAD-ESI-MSn.
Identification of phenolic antioxidants and bioactives of pomegranate seeds following juice extraction using HPLC-DAD-ESI-MSn.
Food chemistry (2016-12-17)
Priyatharini Ambigaipalan, Adriano Costa de Camargo, Fereidoon Shahidi
ABSTRACT
Phenolics from free and hydrolyzed fractions of pomegranate juice (PJ) and seeds (PS) were evaluated. In general, total phenolic contents and scavenging of ABTS+, DPPH and hydroxyl radicals, as well as metal chelation of the soluble fraction from PS, were higher than those for PJ. Insoluble-bound phenolics from PS accounted for up to 27% of total scavenging capacity (free+esterified+insoluble-bound). Phenolic acids (13), monomeric flavonoids (8), hydrolysable tannins (12), proanthocyanidin (1) and anthocyanins (12) were tentatively characterized using HPLC-DAD-ESI-MSn. Several compounds were identified for the first time in PJ or PS. The inhibition of DNA damage (induced by hydroxyl and peroxyl radicals), copper-induced LDL-cholesterol peroxidation, as well as alpha-glucosidase and lipase activities were demonstrated, therefore supporting the potential exploitation of PJ and PS as sources of bioactive compounds.
Sigma-Aldrich
4-Nitrophenyl octanoate, ≥90.0% (GC)
Pricing and availability is not currently available.