HPLC Analysis of Chlorocresol on Chromolith® HighResolution RP-18 endcapped
CONDITIONS
column
Chromolith® HighResolution RP-18 endcapped, 150 x 4.6 mm (1.52023.0001)
mobile phase
[A] water; [B] acetonitrile; (55:45, A:B)
flow rate
1 mL/min
pressure
667 psi (46 Bar)
column temp.
30 °C
detector
240 nm cell: 10 μL
injection
10 μL
sample
Dissolve 10 mg of chlorocresol standard with 100 mL of diluent
Description
Legal Information
Chromolith is a registered trademark of Merck KGaA, Darmstadt, Germany