Merck
HPLC Analysis of Bromelain on TSKgel® BioAssist® S vs. Competitive S Column
HPLC Analysis of Bromelain on TSKgel® BioAssist® S vs. Competitive S Column application for HPLC
Materials
related product
Product No.
Description
Pricing
Bromelain from pineapple stem
≥3 units/mg protein
TSKgel® BioAssist® S HPLC Column
phase sulfonic acid, L × I.D. 5 cm × 4.6 mm, 7 μm particle size, PEEK hardware
TSKgel® BioAssist® S HPLC Column
phase sulfonic acid, L × I.D. 10 cm × 10 mm, 13 μm particle size, PEEK hardware
used together
Product No.
Description
Pricing
Sodium chloride
Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
CONDITIONS
column
TSKgel Bio Assist S, 5 cm x 4.6 mm I.D., 7 μm particles (819686)
column
Competitive S-type column, 5 cm x 5 mm I.D.
mobile phase
[A] 20 mM sodium phophate buffer, pH 7.0; [B] 20 mM sodium phophate buffer, 0.5 M NaCl, pH 7.0
gradient
0 to 100% B in 20 min (0 to 100% B in 30 min for competitive column)
flow rate
0.8 mL/min (1.0 mL/min for competitive column)
column temp.
25 °C
detector
UV, 280 nm
sample
crude bromelain, 0.01 μg/mL
Description
Other Notes
Data provided by Tosoh Bioscience LLC.
Legal Information
BioAssist is a registered trademark of Tosoh Corporation
TSKgel is a registered trademark of Tosoh Corporation