Merck
跳至
ESYNTH018
SynLectro® Graphite Electrode
size 3 mm × 20 mm × 60 mm
登录并加购,请在购物车里查看协议价、货期和发货地。
选择规格
变更视图
规格/货号是否有货价格
1 ea
请联系客服了解存货情况
$340.00
关于此项目
NACRES:
NB.61
UNSPSC Code:
81141502
eCl@ss:
32039090
$340.00
请联系客服了解存货情况
​
申请定制订单
技术服务
需要帮助?我们经验丰富的科学家团队随时乐意为您服务。
让我们为您提供帮助
product line
SynLectro®
size
3 mm × 20 mm × 60 mm
density
1.89 g/cm3 at 25 °C
General description
SynLectro® Graphite Electrode is a 3x20x60 mm electrode designed for use with the SynLectro® Electrolysis Platform for synthetic electrochemistry.
SynLectro® Electrolysis Platform
The SynLectro® Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Legal Information
SynLectro is a registered trademark of Merck KGaA, Darmstadt, Germany
比较类似商品
查看完整比较结果
显示差异
1 of 1
此商品
ESYNTH013ESYNTH011ESYNTH017
description
size 3 mm × 20 mm × 60 mm
description
size 3 mm × 20 mm × 60 mm
description
size 3 mm × 20 mm × 60 mm
description
size 3 mm × 20 mm × 60 mm
size
3 mm × 20 mm × 60 mm
size
3 mm × 20 mm × 60 mm
size
3 mm × 20 mm × 60 mm
size
3 mm × 20 mm × 60 mm
density
1.89 g/cm3 at 25 °C
density
-
density
-
density
-
product line
SynLectro®
product line
SynLectro®
product line
SynLectro®
product line
SynLectro®
历史批次信息供参考:
分析证书(COA)
访问文档库
Questions
  1. 안녕하세요. 줄히팅 60V, 20A 조건에서 기판으로 사용하려고 하는데 적당한 제품인가요?
    1 answer
    1. This product has not been validated for use as a circuit board under the specified conditions of 60V and 20A. It is designed for use with the SynLectro™ Electrolysis Platform for synthetic electrochemistry.