Merck
ESYNTH022
Sigma-Aldrich
SynLectro Graphite Felt Electrode
size 3 mm × 20 mm × 60 mm
Slide 1 of 1
登录查看公司和协议定价
选择尺寸
1 EACH
$173.00
$173.00
请联系客服了解存货情况
获取大包装报价
选择尺寸
变更视图
1 EACH
$173.00
About This Item
分類程式碼代碼:
81141502
NACRES:
NA.06
跳至
技术服务
需要帮助?我们经验丰富的科学家团队随时乐意为您服务。
让我们为您提供帮助
$173.00
请联系客服了解存货情况
获取大包装报价
技术服务
需要帮助?我们经验丰富的科学家团队随时乐意为您服务。
让我们为您提供帮助
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
ESYNTH018ESYNTH019ESYNTH021
size
3 mm × 20 mm × 60 mm
size
3 mm × 20 mm × 60 mm
size
3 mm × 20 mm × 60 mm
size
3 mm × 20 mm × 60 mm
一般說明
SynLectro Graphite Felt Electrode is a 3x20x60 mm electrode designed for use with the SynLectro Electrolysis Platform for synthetic electrochemistry.
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
法律資訊
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
历史批次信息供参考:
分析证书(COA)