Merck
All Photos(2)

GF38160468

foil, thickness 0.015 mm, 6 mm diameter, purity 99.9%

Synonym(s):
MG000180
Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
MDL编号:
PubChem化学物质编号:
NACRES:
NA.23

蒸汽压

1 mmHg ( 621 °C)

测定

99.9%

形式

foil

自燃温度

950 °F

manufacturer/tradename

Goodfellow 381-604-68

电阻率

4.46 μΩ-cm, 20°C

直径

6 mm
6 mm

厚度

0.015 mm

bp

1090 °C (lit.)

mp

648 °C (lit.)

密度

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

一般描述

For updated SDS information please visit www.goodfellow.com

法律信息

Product of Goodfellow

储存分类代码

13 - Non Combustible Solids

WGK

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service