GF81376038

foil, 15mm disks, thickness 0.125mm, as rolled, 99.99+%

别名:
线性分子式:
Ag
CAS号:
分子量:
107.87
MDL编号:
PubChem化学物质编号:

测定

99.99+ %

形式

foil

manufacturer/tradename

Goodfellow 813-760-38

电阻率

1.59 μΩ-cm, 20°C

直径× 厚度

15 mm × 0.125 mm

bp

2212 °C (lit.)

mp

960 °C (lit.)

密度

10.49 g/cm3 (lit.)

SMILES string

[Ag]

InChI

1S/Ag

InChI key

BQCADISMDOOEFD-UHFFFAOYSA-N

一般描述

For updated SDS information please visit www.goodfellow.com.

法律信息

Product of Goodfellow

storage_class_code

13 - Non Combustible Solids

WGK Germany

WGK 3

闪点(F)

Not applicable

闪点(C)

Not applicable

分析证书

原产地证书 (CofO)

Clinical evaluation of C-Film, a vaginal contraceptive.
O Frankman et al.
The Journal of international medical research, 3(4), 292-296 (1975-01-01)
M Dubiel et al.
Journal of synchrotron radiation, 8(Pt 2), 539-541 (2001-08-22)
The thermal expansion behaviour of silver fcc has been described by an anharmonic Einstein model using EXAFS data in the temperature range between 10 and 300 K. The linear expansion coefficient of a bulk silver foil agrees well with X-ray...
A Rupérez et al.
Talanta, 44(2), 213-220 (1997-02-01)
Surface-enhanced Raman scattering (SERS) spectrometry of the diuretic drug triamterene is discussed. The SERS-active substrate used is a silver foil etched with nitric acid. The influence of solvent and sample doping method on sensivity, intercept and shape of the calibration...
H H Wiebe et al.
Plant physiology, 59(2), 256-258 (1977-02-01)
Water potential was monitored at nine locations along single maize (Zea mays L.) leaf blades with aluminum block in situ thermocouple hygrometers. Water potential showed a continuous decrease toward the tip, with a 2- to 4-bar difference between leaf base...
IX. Note on Silver Foil in Surgery.
J S Lewis
Annals of surgery, 50(4), 793-796 (1909-10-01)

我们的科学家团队拥有各种研究领域经验,包括生命科学、材料科学、化学合成、色谱、分析及许多其他领域.

联系技术服务部门