Merck
All Photos(2)

GF92442001

insulated wire, 0.2m, conductor diameter 0.025mm, insulation thickness 0.002mm, PTFE (polytetrafluoroethylene) insulation, 99.99%

Synonym(s):
Empirical Formula (Hill Notation):
Au
CAS Number:
Molecular Weight:
196.97
MDL编号:
PubChem化学物质编号:

测定

≥99.99%

形式

wire

manufacturer/tradename

Goodfellow 924-420-01

电阻率

2.05 μΩ-cm, 0°C

bp

2808 °C (lit.)

mp

1063 °C (lit.)

密度

19.3 g/mL at 25 °C (lit.)

SMILES string

[Au]

InChI

1S/Au

InChI key

PCHJSUWPFVWCPO-UHFFFAOYSA-N

Related Categories

一般描述

For updated SDS information please visit www.goodfellow.com.

法律信息

Product of Goodfellow

储存分类代码

11 - Combustible Solids

WGK

nwg

闪点(F)

Not applicable

闪点(C)

Not applicable

Certificate of Analysis

Enter Lot Number to search for Certificate of Analysis (COA).

Certificate of Origin

Enter Lot Number to search for Certificate of Origin (COO).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service