Merck
AXYPCRAS600
铝密封膜,用于热密封和一般密封
size 50 μm, pkg of 5x100sheets/cs
登录to View Organizational & Contract Pricing
Select a Size
别名:
96 multiwell plate, 96 well PCR plate, 96 well microplate, 96 well microtiter plate, 96 well plate
NACRES:
NB.22
物料
aluminum
无菌性
non-sterile
manufacturer/tradename
Axygen Inc PCR-AS-600
Corning
包装
pkg of 5x100sheets/cs
尺寸
50 μm
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
AXYPCRSPAXYPCRTSAXYAM96PCRRD
CyclerSeal sealing film size 60 μm, polypropylene, pkg of 5x100sheets/cs
AXYPCRTS
CyclerSeal密封薄膜
Sealing mat for PCR microplates, size 96 wells, pkg of 5x10mats/cs
AXYAM96PCRRD
密封垫
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-AS-600, Corning
manufacturer/tradename
Axygen Inc PCR-SP, Corning
manufacturer/tradename
Axygen Inc PCR-TS, Corning
manufacturer/tradename
Axygen Inc AM-96-PCR-RD, Corning
packaging
pkg of 5x100sheets/cs
packaging
pkg of 5x100sheets/cs
packaging
pkg of 5x100sheets/cs
packaging
pkg of 5x10mats/cs
size
50 μm
size
80 μm
size
60 μm
size
96 wells
一般描述
  • Axygen®铝封板膜是光敏样品的理想选择。
  • 均匀的粘合剂实现有效的孔密封。
  • 功能强大的密封膜可用于各种应用,包括 PCR、实时 PCR 、ELISA 和细胞培养。
  • 兼容各种试剂和温度。
法律信息
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA