Merck
AXYPCRTS900
CyclerSeal密封薄膜
size 120 μm, polypropylene, pkg of 5x50sheets/cs
登录to View Organizational & Contract Pricing
NACRES:
NB.22
物料
polypropylene
无菌性
non-sterile
manufacturer/tradename
Axygen Inc PCR-TS-900
Corning
包装
pkg of 5x50sheets/cs
尺寸
120 μm
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
AXYHSFTSLAXYPCRTSAXYPCRSPS
vibrant-m
AXYPCRTS900
CyclerSeal密封薄膜
vibrant-m
AXYHSFTSL
CyclerSeal密封薄膜
CyclerSeal sealing film size 60 μm, polypropylene, pkg of 5x100sheets/cs
AXYPCRTS
CyclerSeal密封薄膜
sterility
non-sterile
sterility
non-sterile
sterility
non-sterile
sterility
sterile
manufacturer/tradename
Axygen Inc PCR-TS-900, Corning
manufacturer/tradename
Axygen Inc, Corning
manufacturer/tradename
Axygen Inc PCR-TS, Corning
manufacturer/tradename
Axygen Inc PCR-SP-S, Corning
packaging
pkg of 5x50sheets/cs
packaging
-
packaging
pkg of 5x100sheets/cs
packaging
pkg of 5x100sheets/cs
size
120 μm
size
-
size
60 μm
size
80 μm
分析证书(COA)
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA