Merck
S2896
Sigma-Aldrich
Scalpel handles
No. 3, stainless steel
登录to View Organizational & Contract Pricing
NACRES:
NB.35
物料
stainless steel
长度
5 in.
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
S9147Z511447Z408220
Scalpel handles No. 3, stainless steel
Sigma-Aldrich
S2896
Scalpel handles
Spatulas Micro; flat rounded ends
S9147
Spatulas
Lab spoons stainless steel, L 9 in.
Sigma-Aldrich
Z511447
Lab spoons
Spatulas with wooden handle blade L × W 8 in. × 1 1/8 in.
Z408220
Spatulas with wooden handle
L
5 in.
L
8 in.
L
9 in.
L
-
应用
Scalpel handles have been used to assemble the scalpel blade to it during adrenalectomy in mice.
分析证书(COA)
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA
其他客户在看
Slide 1 of 5
1 of 5
Stainless steel forceps Dissecting
Z168793
不锈钢镊子
Dissecting scissors Mayo, straight, L 5 1/2 in.
Z265985
Dissecting scissors
BRAND® microscope slide cut edges
BR474701
BRAND®显微镜涂片
Stainless steel forceps Medium tip, curved
Z168696
不锈钢镊子
Micro spatulas Curved at one end with knob at the other end, L 5 1/12 in. (140 mm)
Z193208
Micro spatulas