Merck
All Photos(1)
S6440
Nunc® Lab-Tek® II - CC2腔室玻片系统
1 well
Synonym(s):
chamber slides, microscope slides, microscopy slides
NACRES:
NB.22
物料
colorless
polystyrene chamber
无菌性
sterile
特点
CE marked
manufacturer/tradename
Nunc 154739
包装
pkg of carton of 16
宽度 × 长度
25 mm × 75 mm
well area
8.6 cm2
1
适用性
suitable for (cell culture; optimal for culturing fastidious cells (e.g. neurons))
Looking for similar products? Visit Product Comparison Guide
一般描述
Nunc® Lab-Tek II® - CC2腔室载玻片系统
  • 模拟聚赖氨酸的玻片化学涂层生长表面
  • 提供最适于挑剔细胞(如神经元)的结合位点
  • 无需冷藏维持生长表面的温度
  • 浅蓝色磨砂书写区
  • 每个包装里都带有玻片分离器
  • CE认证
法律信息
CC2 is a trademark of Thermo Fisher Scientific or its subsidiaries
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service