Merck
Z740503
smartSlicer Gel Cutting Tool & RaZor
standard
登录to View Organizational & Contract Pricing
别名:
electrophoresis gel cutter, gel knife, gel razor, gel razor replacement, safety cutter, safety razor, western gel cutting
包装
pack of 100 ea
manufacturer/tradename
LevGo Inc 30202
长度 × 高度 × 厚度
25 mm × 19 mm × 0.27 mm
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 2
此商品
TMKIT
manufacturer/tradename
LevGo Inc 30202
manufacturer/tradename
-
L × H × thickness
25 mm × 19 mm × 0.27 mm
L × H × thickness
-
一般描述
At just 0.27 mm (0.011 in.) the US Patent Pending smartSlicer is as thin as a single edge industrial razor blade but much safer - the smartSlicer is unlikely to slice you!
  • The smartSlicer is able to cleanly slice gels* right on the light Box without scratching the surface of the box
  • The smartSlicer can also be used to chop, mince or slice some soft tissue samples
  • The smartSlicer is also adjustable - simply cut it with standard scissors to the width you require
Available in two sizes: our Standard size is the same size as an industrial single edge razor. Our Large smartSlicer is 30% larger for bigger jobs.
smartSlicers are made in the USA of food grade PET and packed 100 per pack.
Lab Safety Managers love the smartSlicer!
*Best for 1% to 2% gels.
法律信息
smartSlicer is a trademark of LevGo Inc.
分析证书(COA)
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA