Merck
Z744071
General Purpose Sealing Film
polypropylene, sterile
登录to View Organizational & Contract Pricing
Select a Size
物料
polypropylene
无菌性
sterile
特点
RNase and DNase free
包装
case of 100 ea
manufacturer/tradename
Excel Scientific GPS-PP-100
参数
-40-120 °C temp. range
正在寻找类似产品? Visit 产品对比指南
比较类似商品
查看完整比较结果
显示差异
1 of 4
此商品
Z369683Z723304Z744073
vibrant-m
Z744071
General Purpose Sealing Film
ThermalSeal® films, classic sterile, ThermalSeal® Film, Sterile, polypropylene, 100 pack
Sigma-Aldrich
Z369683
ThermalSeal® Film
ThermalSeal A™ sealing films with improved adhesion, clear, polypropylene film, non-sterile
Z723304
ThermalSeal A sealing films
vibrant-m
Z744073
EZcap General Purpose FilmStrips
sterility
sterile
sterility
sterile
sterility
non-sterile
sterility
sterile
packaging
case of 100 ea
packaging
pack of 100 ea
packaging
pack of 100
packaging
case of 400 ea
manufacturer/tradename
Excel Scientific GPS-PP-100
manufacturer/tradename
Excel Scientific STR-THER-PLT
manufacturer/tradename
Excel Scientific TSA-100
manufacturer/tradename
Excel Scientific GPS-CS1X8-400
feature
RNase and DNase free
feature
piercable: no
feature
-
feature
RNase and DNase free
parameter
-40-120 °C temp. range
parameter
-
parameter
-
parameter
-40 °C temp. range (120 °C)
一般描述
  • Certified DNase- & RNase- free
  • Designed to be used with both polystyrene and polypropylene plates
  • 2.0 mil clear polypropylene film features good clarity and easy handling
  • Specially formulated adhesive provides an excellent seal to minimize evaporation, reduce cross-contamination and prevent spillage
  • Removes easily and cleanly from plate surfaces
  • Minimizes aerosols upon film removal
  • Features two end-tabs with perforations for ease of handling, application and removal
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Documents related to the products that you have purchased in the past have been gathered in the Document Library for your convenience.
Visit the Document Library
Difficulty Finding Your Product Or Lot/Batch Number?
在网站页面上,产品编号会附带包装尺寸/数量一起显示(例如:T1503-25G)。请确保 在“产品编号”字段中仅输入产品编号 (示例: T1503).
示例
T1503
货号
-
25G
包装规格/数量
其它示例:
705578-5MG-PW
PL860-CGA/SHF-1EA
MMYOMAG-74K-13
1000309185
输入内容 1.000309185)
遇到问题?欢迎随时联系我们技术服务 寻求帮助
批号可以在产品标签上"批“ (Lot或Batch)字后面找到。
Aldrich 产品
  • 如果您查询到的批号为 TO09019TO 等,请输入去除前两位字母的批号:09019TO。
  • 如果您查询到的批号含有填充代码(例如05427ES-021),请输入去除填充代码-021的批号:05427ES。
  • 如果您查询到的批号含有填充代码(例如 STBB0728K9),请输入去除填充代码K9的批号:STBB0728。
未找到您寻找的产品?
部分情况下,可能未在线提供COA。如果搜索不到COA,可在线索取。
索取COA