Merck
All Photos(1)
Z744072
EZcap General Purpose FilmStrips
polypropylene, non-sterile
物料
polypropylene
无菌性
non-sterile
特点
RNase and DNase free
包装
case of 400 ea
manufacturer/tradename
Excel Scientific GP-CS1X8-400
参数
-40 °C temp. range (120 °C)
Looking for similar products? Visit Product Comparison Guide
一般描述
  • Certified DNase- & RNase- free
  • Designed to be used with both polystyrene and polypropylene plates
  • Precision cut to a smaller dimension to seal a single 8-well row of a 96-well plate or an ELISA strip-well plate
  • Each sheet contains 8 FilmStrips on a continuous backing liner for ease of use
  • FilmStrips can be peeled and applied individually or two-at-a-time for increased efficiency
  • FilmStrips can be used to selectively cover one row of a plate to prevent accidental pipetting or to re-seal rows that have been pierced
  • 2.0 mil polypropylene film features good clarity and easy handling
  • Specially formulated adhesive provides an excellent seal to minimize evaporation, reduce cross-contamination and prevent spillage
  • Removes easily and cleanly from plate surfaces
  • Minimizes aerosols upon film removal
  • Features two end-tabs with perforations for ease of handling, application and removal
法律信息
EZcap is a trademark of Excel Scientific, Inc.
FilmStrips is a trademark of Excel Scientific, Inc.
Certificate of Analysis
Enter Lot Number to search for Certificate of Analysis (COA).
Certificate of Origin
Enter Lot Number to search for Certificate of Origin (COO).
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service