SG
ZH
应用
产品
服务
支持
Home
Search Results
镀金显微镜载片
(2)
镀金显微镜载片
Linear Formula:
Au
CAS Number:
7440-57-5
Molecular Weight:
196.97
EC Number:
231-165-9
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
643203
镀金显微镜载片, layer thickness 100 Å, 99.999% (Au)
Expand
View Pricing
643246
镀金显微镜载片, layer thickness 1000 Å, 99.999% (Au)
Expand
View Pricing